Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Endothelin 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113958
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113958 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113958 Supplier Invitrogen™ Supplier No. PA5113958
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MVSVPTTWCS VALALLVALH EGKGQAAATL EQPASSSHAQ GTHLRLRRCS For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 93%.

Endothelins are endothelium-derived vasoconstrictor peptides. Endothelin 2, also known as ET-2, is a protein that in humans is encoded by the EDN2 gene. The secreted peptide (~2kD, 21 amino acids) is produced as a result of endothelin-2 precursor (~20 kD) processing and functions as a ligand for endothelin receptors which upon peptide binding modulates a wide array of cell signaling events. Diseases associated with endothelins include aortic disease and patent ductus arteriosus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Endothelin 2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene EDN2
Gene Accession No. P20800
Gene Alias EDN 2; Edn2; EDN-2; endothelin 2; Endothelin isopeptide 2; endothelin-2; ET2; ET-2; PP ET-2; PPET2; preproendothelin 2; preproendothelin-2; RNEDN2S01; vasoactive intestinal contractor; vasoactive intestinal contractor peptide; VIC
Gene Symbols EDN2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the n terminal region of human EDN2 (aa 1-50).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1907
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.