Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Endothelin A Receptor Polyclonal Antibody, Invitrogen™

Catalog No. PIPA530589
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530589 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA530589 Supplier Thermo Scientific Supplier No. PA530589
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: U87-MG, GL261. Predicted reactivity: Rhesus Monkey (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes the receptor for endothelin-1, a peptide that plays a role in potent and long-lasting vasoconstriction. This receptor associates with guanine-nucleotide-binding (G) proteins, and this coupling activates a phosphatidylinositol-calcium second messenger system. Polymorphisms in this gene have been linked to migraine headache resistance. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Endothelin A Receptor
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.78 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene EDNRA
Gene Accession No. P25101, P26684, Q61614
Gene Alias Ednra; Endor; Endothelin A; endothelin A receptor; endothelin receptor subtype A; endothelin receptor type A; endothelin type A receptor; endothelin-1 receptor; endothelin-1-specific receptor; ETa; ET-A; ETAR; ET-AR; ETA-R; ETRA; G protein coupled receptor; G protein-coupled receptor; Gpcr10; G-protein coupled receptor 10; hET-AR; MFDA; RGD1559432
Gene Symbols EDNRA
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a region within amino acids 363 and 427 of Human Endothelin A Receptor
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13617, 1909, 24326, 373908
Target Species Chicken, Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.