Learn More
Invitrogen™ Endothelin B Receptor Polyclonal Antibody

Description
Percent identity with other species by BLAST analysis: Human, Dog (100%) Gorilla, Gibbon (95%) Monkey (89%) Marmoset (84%).
Specifications
Specifications
| Antigen | Endothelin B Receptor |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide |
| Gene | EDNRB |
| Gene Accession No. | P24530, P56497 |
| Gene Alias | ABCDS; AU022549; Ednra; Ednrb; endothelin B receptor; endothelin receptor non-selective type; endothelin receptor subtype B1; endothelin receptor type B; endothelin-B receptor; ETb; ET-B; Etb receptor; Et-b receptor; ETB1; ETBR; ET-BR; Et-b-r; Etb-svr; ETRB; ET-RB; ETR-b; HSCR; HSCR2; piebald; RP11-318G21.1; Sox10m1; WS4A |
| Gene Symbols | EDNRB |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.