Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Endothelium Monoclonal Antibody (OX-43)
GREENER_CHOICE

Catalog No. PIMA170023
Change view
Click to view available options
Quantity:
250 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA170023 250 μg
PIMA517513 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA170023 Supplier Invitrogen™ Supplier No. MA170023
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

For extended storage, aliquot and store at -20°C or below. Avoid freeze-thaw cycles.

The endothelium is the layer of thin specialized epithelium, comprising a simple squamous layer of cells that line the interior surface of blood vessels, forming an interface between circulating blood in the lumen and the rest of the vessel wall (simple squamous epithelium). Endothelial cells line the entire circulatory system, from the heart to the smallest capillary. These cells reduce friction of the flow of blood.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Endothelium
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone OX-43
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Alias Endothelial cell marker; Endothelial cells; Endothelium Cell Antigen-1; Pan endothelial cells
Host Species Mouse
Immunogen Rat peritoneal macrophages.
Purification Method Protein G
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.