Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Endothelium Monoclonal Antibody (OX-43), FITC
Mouse Monoclonal Antibody
$313.00 - $636.50
Specifications
Antigen | Endothelium |
---|---|
Clone | OX-43 |
Concentration | 0.1 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. |
Applications | Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA517516
|
Invitrogen™
MA517516 |
100 μg |
Each for $313.00
|
|
|||||
PIMA517517
|
Invitrogen™
MA517517 |
500 μg |
Each for $636.50
|
|
|||||
Description
Endothelium Monoclonal Antibody for Flow
The endothelium is the layer of thin specialized epithelium, comprising a simple squamous layer of cells that line the interior surface of blood vessels, forming an interface between circulating blood in the lumen and the rest of the vessel wall (simple squamous epithelium). Endothelial cells line the entire circulatory system, from the heart to the smallest capillary. These cells reduce friction of the flow of blood.Specifications
Endothelium | |
0.1 mg/mL | |
Flow Cytometry | |
FITC | |
Mouse | |
RUO | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
Rat Peritoneal Macrophages. | |
Antibody |
OX-43 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
Monoclonal | |
Liquid | |
IgG1 | |
Rat | |
Endothelial cell marker; Endothelial cells; Endothelium Cell Antigen-1; Pan endothelial cells | |
Primary | |
Protein G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title