Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Endov Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570136
Description
This target displays homology in the following species: Cow: 100%; Dog: 93%; Guinea Pig: 79%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 77%.
ENDOV gene ontology annotations related to this gene include DNA repair.
Specifications
Endov | |
Polyclonal | |
Unconjugated | |
ENDOV | |
A730011L01Rik; Endonuclease V; Endov; hEndoV; inosine-specific endoribonuclease; novel endonuclease V protein; putative endonuclease FLJ39025 homolog | |
Rabbit | |
Affinity chromatography | |
RUO | |
338371 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8C9A2 | |
ENDOV | |
synthetic peptide directed towards the middle region of human Endov (aa 138-187). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction