Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Endoxifen

Catalog No. 370510R
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-051-0R 10 mg
37-055-0R 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 37-051-0R Supplier Tocris Bioscience™ Supplier No. 3705/10
This item is not returnable. View return policy

Potent antiestrogen; ERα ligand

Endoxifen is an estrogen receptor α (ERα) ligand; potent antiestrogen. Metabolite of tamoxifen (Cat. No. 0999). Primary metabolite responsible for the effectiveness of tamoxifen in ER-positive breast cancer.

Specifications

Purity >98%
CAS 110025-28-0
Quantity 10 mg
Product Type Endoxifen
Molecular Weight (g/mol) 373.49
Inhibitors Endoxifen

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.