Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Mouse anti-Human, Clone: ENG/1326, Abnova™

Catalog No. 89034595 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-034-595 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-034-595 Supplier Abnova Corporation Supplier No. MAB14512
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against partial recombinant human ENG.

  • High specificity
  • High reactivity

Specifications

Antigen ENG
Applications Flow Cytometry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone ENG/1326
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant human ENG.
Dilution Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Western Blotting (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
Gene Accession No. P17813
Gene Alias CD105/END/FLJ41744/HHT1/ORW/ORW1
Gene Symbols ENG
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 74-251 of human ENG.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2022
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.