Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Acid-Fast Stain Kit, Modification of the Kinyoun Method

Catalog No. p-3201183
Click to view available options
Stain Type:
Brilliant Green
Loeffler's Methylene Blue
This item is not returnable. View return policy
This item is not returnable. View return policy

This Acid-Fast Stain Kit, Modification of the Kinyoun Method, is ideal for staining acid-fast bacteria. Use with Acid-fast stain kit.

  • Modification of the Kinyoun Method
  • Acid-fast bacilli stain red; all other bacteria stain blue with Sol. III, green with Sol. IIIA.
  • Acid Fast Stain Kit
  • For in vitro diagnostic use
  • 1 Kit

Specifications

Type Acid Fast Stain
Quantity 1 Ea.
Includes 250 mL carbol fuchsin, 250 mL acid alcohol and 250 mL Loeffler's methylene blue
For Use With (Application) In vitro diagnostic use
Color Dark Red
Packaging Type Kit
Stain Type Loeffler's Methylene Blue
Form Liquid

For in vitro diagnostic use only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.