Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Bennhold's Congo Red Stain Kit

Catalog No. ES9210
Click to view available options
Type:
Bennhold Congo Red Stain Kit
Solution I Congo Red
Solution II Lithium Carbonate
Solution III Ethanol
Solution IV Harris Alum Hematoxylin

Catalog No. ES9210

Supplier: ENG Scientific 9210

Only null left
Add to Cart
Add to Cart

Includes

Made from certified dyes (when applicable). Kit includes:

  • Solution I - Congo Red
  • Solution II - Lithium Carbonate (Saturated Aqueous)
  • Solution III - Ethanol (80%)
  • Solution IV - Harris' Alum Hematoxylin

Permanent stain for amyloid.

  • Permanent stain for amyloid
  • Made from certified dyes (when applicable)
  • Kit includes:
    • Solution I - Congo Red
    • Solution II - Lithium Carbonate (Saturated Aqueous)
    • Solution III - Ethanol (80%)
    • Solution IV - Harris' Alum Hematoxylin
  • For in vitro diagnostic use

Specifications

Type Solution I Congo Red
For Use With (Application) Permanent stain for amyloid
Packaging Type Bottles
Quantity 2 x 250 mL

For in vitro diagnostic use only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.