Learn More
ENG Scientific Bennhold's Congo Red Stain Kit
Catalog No. es9200
Click to view available options
Type:
Bennhold Congo Red Stain Kit
Solution I Congo Red
Solution II Lithium Carbonate
Solution III Ethanol
Solution IV Harris Alum Hematoxylin
Includes
Made from certified dyes (when applicable). Kit includes:
  • Solution I - Congo Red
  • Solution II - Lithium Carbonate (Saturated Aqueous)
  • Solution III - Ethanol (80%)
  • Solution IV - Harris' Alum Hematoxylin
Permanent stain for amyloid.
  • Permanent stain for amyloid
  • Made from certified dyes (when applicable)
  • Kit includes:
    • Solution I - Congo Red
    • Solution II - Lithium Carbonate (Saturated Aqueous)
    • Solution III - Ethanol (80%)
    • Solution IV - Harris' Alum Hematoxylin
  • For in vitro diagnostic use
Specifications
Type Bennhold Congo Red Stain Kit
For Use With (Application) Permanent stain for amyloid
Includes Congo Red, Lithium Carbonate, Ethanol, Hematoxylin
Packaging Type 1 Kit (Bottles)
Quantity 4 x 250 mL
For in vitro diagnostic use only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.