Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Bismarck Brown with Methyl Green Stain Kit

Catalog No. p-3256219
Click to view available options
Quantity:
2 x 250 mL
2 x 950 mL
Type:
Bismarck Brown with Methyl Green Stain Kit
Solution I Aqueous Bismarck Brown
Solution II Aqueous Methyl Green
This item is not returnable. View return policy
This item is not returnable. View return policy

For intestinal (calciform cells), tracheal, and embryonic tissue.

  • Kit includes Solution I - 1% Aqueous Bismarck Brown and Solution II - 0.5% Aqueous Methyl Green
  • Made from certified dyes
  • For in vitro diagnostics use only
  • Used for intestinal (calciform cells), tracheal and embryonic tissue

Specifications

Type Bismarck Brown with Methyl Green Stain Kit
For Use With (Application) For intestinal (calciform cells), tracheal and embryonic tissue
Includes 1% Aqueous Bismarck Brown, 0.5% Aqueous Methyl Green
Packaging Type Kit (Bottles)
Quantity 2 x 250 mL

For in vitro diagnostic use only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.