Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ENG Scientific Calcofluor White Stain, Certified
Click to view available options
Quantity:
100 mL
2 x 100 mL
Description
- Fungi elements fluoresce blue-white or green depending on filter system used
- A G-365 excitation filter and an LP 420 barrier filter produce light of blue-white wavelength
- A K530 excitation filter and a BG 12 barrier filter produce light of green wavelength
- Calcofluor White Stain
- Includes Evans blue
- For in vitro diagnostic use
Specifications
Specifications
| Type | Calcofluor White Stain |
| Quantity | 2 x 100 mL |
| Includes | Evans blue |
| Format | Bottles |
| For Use With (Application) | In vitro diagnostic use |
For in vitro diagnostic use only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction