Learn More
ENG Scientific Fecal Leukocyte Stain (Loeffler' Methylene Blue)
Catalog No. p-3605860
Click to view available options
Quantity:
1 x 100 mL
2 x 100 mL
Fecal Leukocyte Stain (Loeffler'sMethylene Blue) is for microscopic identification and counting of fecal leukocytes. Smears are alcohol-ether fixed.
  • Contents: Ethanol, Methylene Blue
  • This solution is made from certified dyes.
Specifications
Quantity 1 x 100 mL
For Use With (Application) For microscopic identification and counting of fecal leukocytes
Staining Time 2 to 3 min.
Packaging Bottle
For in vitro diagnostic use only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.