Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Giemsa Stain

Catalog No. es3001
Click to view available options
Quantity:
100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Use this hematology stain for manual staining techniques. Dilute for a counterstain or use as straight stain.

  • Contains methanol
  • Stain or counterstain

Specifications

Quantity 100 mL
For Use With (Application) For in-vitro diagnostic use, manual staining techniques
Packaging Bottle

For in vitro diagnostic use only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.