Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ENG Scientific Gram Stain Kit with Carbol Fuchsin Counterstain
Click to view available options
Quantity:
1 x 250 mL
4 x 250 mL
Type:
Acetone-Alcohol
Carbol Fuchsin
Crystal Violet
Gram Iodine
Gram Stain Kit
Packaging Type:
Bottle
Complete Kit (Bottles)
Description
- Contains crystal violet, iodine, potassium oxalate, ethanol, acetone, safranin, basic fuchsin, phenol
- Gram-positive bacteria, many fungi and fibrin stain blue-black to violet
- Gram-negative bacteria stain red
- Cytoplasm stains pink
Specifications
Specifications
Type | Gram Iodine |
Quantity | 1 x 250 mL |
For Use With (Application) | In vitro diagnostic use |
Packaging Type | Bottle |
Stain Type | Gram Stain |
For in vitro diagnostic use only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction