Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ENG Scientific Gram Stain Kit with Safranin Counterstain
Click to view available options
Type:
Crystal Violet
Crystal Violet, Safranin
Gram Iodine
Safranin
For Use With (Application):
Bacteria in tissue
Gram staining
In vitro diagnostic use
Description
- Contains crystal violet, iodine, potassium oxalate, ethanol, acetone, safranin, basic fuchsin, phenol
- Gram-positive bacteria, many fungi and fibrin stain blue-black to violet
- Gram-negative bacteria stain red
- Cytoplasm stains pink
Specifications
Specifications
| Type | Crystal Violet, Safranin |
| Quantity | 4 x 250 mL |
| For Use With (Application) | In vitro diagnostic use |
| Packaging Type | Complete Kit (Bottles) |
| Stain Type | Gram Stain |
For in vitro diagnostic use only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction