Learn More
ENG Scientific Heinz Body Stain
Catalog No. p-3274902
Click to view available options
Quantity:
100 mL
2 x 100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy
Use this helpful Heinz Body Stain to recognize hemolytic anemia due to certain drugs and in detecting red cells of the primaquine-sensitive type by detecting Heinz bodies.
  • Heinz bodies are best stained supravitally
  • Includes Methyl Violet, Saline (NaCl)
  • Made from certified dyes
Specifications
Quantity 100 mL
Type Heinz Body Stain
For Use With (Application) Hematology Staining
Packaging Bottle
For in vitro diagnostic use only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.