Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Herxheimer's Sudan IV Stain Kit

Catalog No. p-3256227
Click to view available options
Quantity:
2 x 250 mL
3 x 250 mL
Type:
Herxheimer Sudan IV Stain Kit
Solution I Sudan IV (Scarlet Red)
Solution II Ethanol
Solution III Mayer's Acid Alum Hematoxylin

More soluble in fat than in alcohol so fat absorbs the stain.

  • More soluble in fat than in alcohol so fat absorbs the stain
  • Kit includes:
    • Solution I - Sudan IV (Scarlet Red)
    • Solution II - Ethanol (70%)
    • Solution III - Mayer's Acid Alum Hematoxylin
  • Used to depict nuclei, fat, normal myelin and fatty acids
  • Made from certified dyes (when applicable)

Specifications

Type Solution I Sudan IV (Scarlet Red)
For Use With (Application) Depicts nuclei, fat, normal myelin and fatty acids
Packaging Type Bottles
Quantity 2 x 250 mL

For in vitro diagnostic use only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.