Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Macchiavello Stain Kit

Catalog No. p-4717516
Click to view available options
Quantity:
4 x 100 mL
4 x 250 mL

Differential staining procedure based on the affinity of cationic (basic) dyes for rickettsiae and chlamydiae.

  • Rickettsiae and chlamydiae stain red, tissue cells stain blue
  • For Macchiavello Stain Kit
  • Includes 250 mL of Solution I - Basic Fuchsin, Solution II - 0.5% Citric Acid, Solution III - Methylene Blue, and Solution IIIA - Malachite Green

Specifications

Type Macchiavello Stain Kit
Quantity 4 x 100 mL
Includes 100 mL each of Solutions I, II, III and IIIA
For Use With (Application) Rickettsiae and Chlamydiae
Composition Basic Fuchsin, Citric Acid, Methylene Blue, Malachite Green

For in vitro diagnostic use only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.