Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Maldonado's Method Stain Kit

Catalog No. p-3256228
Click to view available options
Quantity:
2 x 250 mL
5 x 250 mL
Reagent Type:
Azure II (0.05%)
Complete Kit
Phloxine (1%)
Phosphotungstic Acid (3%)
Weigert's Hematoxylin
Weigert's Iron Chloride

For pancreatic islet cells.

  • Kit includes:
    • Solution I - Phloxine (1%)
    • Solution II - Phosphotungstic Acid (3%)
    • Solution III - Azure II (0.05%)
    • Solution IVA - Weigert's Hematoxylin
    • Solution IVB - Weigert's Iron Chloride
  • Maldonado's Method Stain Kit
  • Used with Pancreatic islet cells

Specifications

Content And Storage Solution I - Phloxine (1%), Solution II - Phosphotungstic Acid (3%), Solution III - Azure II (0.05%), Solution IVA - Weigert's Hematoxylin, and Solution IVB - Weigert's Iron Chloride
Packaging Type Complete Kit (Bottles)
Product Type Staining Reagent
Quantity 5 x 250 mL
For Use With (Application) Pancreatic islet Cells
Reagent Type Complete Kit

For in vitro diagnostic use only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.