Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Manual Method Stain Buffer Solution

Catalog No. p-3605896
Click to view available options
Quantity:
250 mL
3800 mL (1 gal.)
950 mL

Manual Method Stain Buffer Solution is useful for examining bloody specimen material or blood culture supernatant fluid.

  • This solution, using a flooding technique, will stain all cellular elements without precipitate or artifactural changes.
  • Staining characteristics are similar to H-PACK.
  • Buffer is applied immediately after stain, reducing overall staining time.
  • Convenient flip-top cap for 250 mL and 950 mL sizes.
  • IN THE EVENT OF COLD WEATHER, allow solution to return to room temperature and SHAKE WELL BEFORE USE.
  • Contents: Wright Stain, Giemsa Stain, Methanol, Buffers, Stabilizers.
  • These solutions are made from certified dyes (when applicable).

Specifications

Quantity 250 mL
Storage Requirements Room Temperature
For Use With (Application) For examining Bloody specimen material or Blood Culture supernatant fluid, in-vitro diagnostic use
pH 6.73 to 6.77
Packaging Bottle

For in vitro diagnostic use only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.