Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ENG Scientific Manual Method Stain, Wright Stain and Matching Buffer
Click to view available options
Quantity:
2 x 250 mL
2 x 3800 mL (1 gal.)
2 x 950 mL
Description
- Solution uses flooding technique
- Staining time: 5 to 7 minutes
- Buffer is applied immediately after stain, reducing overall staining time
- Convenient flip-top cap for 250 mL and 950 mL sizes
- In the event of cold weather, allow solutions to return to room temperature
Specifications
Specifications
| Quantity | 2 x 250 mL |
| Storage Requirements | Room Temperature |
| For Use With (Application) | For staining of cellular elements without precipitate or artifactural changes, in-vitro diagnostic use |
| Staining Time | 5 to 7 min. |
For in vitro diagnostic use only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction