Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific May-Grunwald Stain Kit May-Grunwald Stain Solution I

Catalog No. p-3605903
Click to view available options
Quantity:
2 x 100 mL
2 x 250 mL

The May-Grunwald Stain Kit May-Grunwald Stain Solution I is used for staining bone marrow cells.

  • Solution I May Grunwald Stain
  • Solution II Stock Giemsa
  • Contents: May-Grunwald Stain, Giemsa Stain, Methanol
  • Solutions made from certified dyes (when applicable)

Specifications

Quantity 2 x 250 mL
For Use With (Application) For use in May-Grunwald staining for bone marrow cells
Staining Time 3 min.
Packaging Bottles

For in vitro diagnostic use only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.