Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Mayer's Mucicarmine Stain

Catalog No. p-3274908
Click to view available options
Quantity:
100 mL
250 mL

Mayer's Mucicarmine Stain is designed to show mucin derived from epithelial cells (Bronchi and Colon).

  • Includes Carmine, Aluminum Chloride
  • Solution made from certified dyes

Specifications

Content And Storage Carmine, Aluminum Chloride
Packaging Type Bottle
Product Type Staining Reagent
Quantity 100 mL
For Use With (Application) In vitro diagnostic use only
Test Time 15 min.

For in vitro diagnostic use only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.