Learn More
ENG Scientific Nile Blue A Stain (Basic Blue 12, Nile Blue Sulfate, Niblau BX)
Click to view available options
Quantity:
250 mL
2 x 250 mL
Description
- Nile Blue A Stain
- Nile Blue A, Sulfuric Acid
- Made from certified dyes
- For in vitro diagnostics use only
- For the differentiation of melanins and lipofuscins
Specifications
Specifications
Type | Nile Blue A Stain |
For Use With (Application) | Differentiation of melanins and lipofuscins |
Includes | Nile Blue A, Sulfuric Acid |
Packaging Type | Bottle |
Quantity | 250 mL |
For in vitro diagnostic use only.