Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ENG Scientific Platelet Diluent
Click to view available options
Quantity:
1 x 100 mL
2 x 100 mL
Description
- Contains Ammonium Oxalate
- For in vitro use only
Specifications
Specifications
Quantity | 2 x 100 mL |
For Use With (Application) | For platelet counts, in-vitro diagnostic use only |
Staining Time | 15 to 20 min. |
Packaging | Bottles |
For in vitro diagnostic use only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction