Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ENG Scientific Solution II - Acid Alcohol
Click to view available options
Quantity:
1 x 3800 mL (1 gal.)
2 x 250 mL
2 x 950 mL
For Use With (Equipment):
Acid-fast Stain Kit (Modified Kinyoun Method)
Cryptosporidium Stain Kit (Modified Kinyoun Method)
Ziehl-Neelsen Stain Kit
Description
- Solution II - Acid Alcohol
Specifications
Specifications
Description | Prepared from fresh material or preserved in 10% buffered formalin. |
Type | Solution II - Acid Alcohol |
For Use With (Application) | Differentiates Cryptosporidium oocysts from yeast cells |
For Use With (Equipment) | Cryptosporidium Stain Kit (Modified Kinyoun Method) |
Packaging Type | Bottle |
Quantity | 2 x 250 mL |
For in vitro diagnostic use only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction