Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Solution IIIA - Brilliant Green

Catalog No. p-4717556
Click to view available options
Quantity:
1 x 3800 mL (1 gal.)
2 x 250 mL
2 x 950 mL

Solution IIIA - Brilliant Green is ideal for staining acid-fast bacteria.

  • Use with Acid-fast stain kit.
  • Modification of the Kinyoun Method.

Specifications

Type Solution IIIA - Brilliant Green
For Use With (Application) Acid-fast stain kit
Quantity 2 x 250 mL

For in vitro diagnostic use only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.