Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Solutions IA and IB

Catalog No. p-4717575
Click to view available options
Quantity:
Solution IA: 1 x 3800 mL, Solution IB: 1 x 450 mL
Solution IA: 2 x 250 mL, Solution IB: 2 x 15 mL

Solutions IA and IB for staining of intestinal protozoa.

  • Use with Trichrome Stain Kit (For Intestinal Protozoa)
  • Kit components: Solutions IA - Schaudinn Solution and IB - Glacial Acetic Acid
  • Not to be confused with Gomori's Trichrome Stain Used for Histology

Specifications

Type Solution IA and IB (Schaudinn Solution and Glacial Acetic Acid)
For Use With (Application) Ideal for staining of intestinal protozoa
Quantity Solution IA: 1 x 3800 mL, Solution IB: 1 x 450 mL

For in vitro diagnostic use only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.