Learn More
ENG Scientific Stat-Quick Wright and Wright Giemsa Stain Kits
Catalog No. p-3605945
Click to view available options
Quantity:
2 x 3800 mL (1 gal.)
2 x 950 mL
Type:
Wright Giemsa Stain and Matching Buffer
Wright Stain and Matching Buffer
Utilized in both stat and routine laboratories. For hematology staining. Stat Quick Stain (Wright or Wright Giemsa) and Buffers.
  • Only two solutions needed to obtain excellent staining
  • No fixation required with this stain
  • With matching buffer solution
  • Matching buffer solution allows uniform staining of all cellular elements without precipitate or artifactual changes
  • Stain placed in staining container should be mixed periodically
  • During cold weather, allow solution to return to room temperature and shake well before use
  • Used in both stat and routine laboratories
Specifications
Quantity 2 x 950 mL
Type Wright Stain and Matching Buffer
Packaging Kit
For in vitro diagnostic use only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.