Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ENG Scientific Sudan Black B Stain Kit
Click to view available options
Quantity:
2 x 250 mL
2 x 950 mL
4 x 250 mL
Type:
Solution I Sudan Black B
Solution II Buffer
Solution III Alcohol (Ethanol, 70%)
Solution IV Harris Alum Hematoxylin
Sudan Black B Stain Kit
Description
- Contains Sudan Black B, ethanol, buffers, stabilizers, hematoxylin, potassium alum, mercuric oxide
Specifications
Specifications
| Type | Solution I Sudan Black B |
| For Use With (Application) | For differential diagnosis of acute granulocytic leukemia, acute lymphocytic and acute myelo-monocytic leukemia |
| Packaging Type | Bottles |
| Quantity | 2 x 950 mL |
For in vitro diagnostic use only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction