Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ENG Scientific Traxstain May-Grunwald Giemsa
Click to view available options
Quantity:
1 x 3800 mL
Description
- Traxstain May-Grunwald Giemsa
- For EM Sciences Midas™ slide stainers, Geometric Data Corporation Hemastainer™ or any other instrument employing a dip stain technique
Specifications
Specifications
Quantity | 1 x 3800 mL |
Storage Requirements | Room Temperature |
For Use With (Equipment) | For EM Sciences Midas™ slide stainers, Geometric Data Corporation Hemastainer™ or any other instrument employing a dip stain technique |
Packaging | Cubetainer |
For in vitro diagnostic use only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction