Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ENG Scientific Traxstain Wright Giemsa Stain
Click to view available options
Quantity:
1 x 3800 mL (1 gal.)
2 x 950 mL
Description
- In the event of cold weather, allow solution to return to room temperature and shake well before use.
- Contents: Wright Stain, Giemsa Stain, May-Grunwald Stain, Methanol, Buffers, Stabilizers.
- These solutions are made from certified dyes (when applicable).
Specifications
Specifications
Quantity | 1 x 3800 mL (1 gal.) |
Storage Requirements | Room Temperature |
Type | Wright Giemsa Stain |
For Use With (Equipment) | For instruments using a dip stain technique |
Packaging | Cubetainer |
For in vitro diagnostic use only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction