Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Urinary Sediment Stain

Catalog No. es3150
Click to view available options
Quantity:
1 x 100 mL
2 x 100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

A quick staining procedure by which a drop of stain solution is directly added to urine sediment.

  • Modification of Sternheimer and Malbin
  • Contents: Phloxine, Propylene Glycol, Sodium Carbonate Phloxine, Propylene Glycol, Sodium Carbonate
  • Solution made from certified dyes

Specifications

Type Urinary Sediment Stain
For Use With (Application) Visualize neutrophils, glitter and squamous cells, bacteria and hyaline, granular and cellular casts
Includes Crystal violet, ammonium oxalate, ethanol, safranin
Packaging Type Bottles
Quantity 2 x 100 mL

For in vitro diagnostic use only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.