Learn More
ENG Scientific WBC Diluting Fluid
Catalog No. p-3274925
Click to view available options
Quantity:
1 x 100 mL
2 x 100 mL
Catalog No. ES5401
Supplier: ENG Scientific 5401
Only null left
Add to Cart
Add to Cart
WBC Diluting Fluid is a modified solution which hemolyzes RBC's and intensifies the cellular outline of WBC's.
  • Contents: Glacial Acetic Acid
Specifications
Quantity 1 x 100 mL
Type WBC Diluting Fluid
For Use With (Application) Hemolyze RBCs and intensify cellular outline of WBCs in blood sample
For in vitro diagnostic use only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.