Learn More
ENG Scientific Wright Giemsa Quick Stain Kit
Catalog No. p-4883101
Click to view available options
Quantity:
3 x 3800 mL (1 gal.)
3 x 475 mL
This item is not returnable. View return policy
This item is not returnable. View return policy
For excellent staining results in one minute without precipitate. Alcoholic quick stain technique. No fixation necessary. For Quick Stain Kit (Wright or Wright Giemsa).
  • Less time in Solution III provides a more alkaline (blue) stain
  • Increase time for a more acidic (red) stain
  • For excellent staining results in one minute
Specifications
Quantity 3 x 475 mL
For Use With (Application) For excellent staining results in one min.
Packaging Complete Kit (Bottles)
For in vitro diagnostic use only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.