Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ENG2A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143401
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143401 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143401 Supplier Invitrogen™ Supplier No. PA5143401
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

Predicted to enable DNA-binding transcription factor activity, RNA polymerase II-specific and RNA polymerase II cis-regulatory region sequence-specific DNA binding activity Acts upstream of or within apoptotic process involved in morphogenesis; cell fate specification; and nervous system development Predicted to be active in nucleus Is expressed in several structures, including adaxial cell; central nervous system; muscle; neural plate; and pharyngeal arch Human ortholog(s) of this gene implicated in autistic disorder Orthologous to human EN2 (engrailed homeobox 2).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ENG2A
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene Alias eng2; ENG-2; eng2a; engrailed homeobox 2a; Zf-en; ZF-EN-2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of zebrafish Eng2a. The exact sequence is proprietary.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.