Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Engineer and Explore Materials for Prosthetics Kit
GREENER_CHOICESCI_ED

Catalog No. S27127 Shop All Aldon Products
Change view
Click to view available options
Class Size:
15 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S27127 15 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: CHOKING HAZARD, Children under 8 yrs. can choke or suffocate on uninflated or broken balloons. Adult supervision required. Keep uninflated balloons from children. Discard broken balloons at once.
Catalog No. S27127 Supplier Innovating Science™ Supplier No. IS3404
Add to Cart
Add to Cart
WARNING: CHOKING HAZARD, Children under 8 yrs. can choke or suffocate on uninflated or broken balloons. Adult supervision required. Keep uninflated balloons from children. Discard broken balloons at once.

Innovating Science™ Engineer and Explore Materials for Prosthetics Kit investigates materials that biomedical engineers use when creating prosthetic devices. Students identify the physical properties of various metals, ceramics, and polymers to determine which are most suitable for use.

  • In this lab, students will explore the materials that biomedical engineers use when creating prosthetic devices
  • They will first investigate the physical properties of various metals, ceramics, and polymers to understand how they differ
  • Then, they will determine which materials would be most suitable for use in a hip replacement prosthetic
  • Students will then engineer their own prosthetic arm that meets certain design requirements and mimics the functionality of a real hand
  • Kit contains enough materials for 15 groups
  • Teacher’s Manual and Student Study Guide copy masters are included

Specifications

Certifications/Compliance Non-regulated
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Educational - Biomedical Engineering
Includes 15 Bone Pieces, 30 Aluminum Strips, 30 Cobalt Pieces, 30 Brass Foil Strips, 30 Zinc Pieces, 30 Ceramic Tile Pieces, 72 Glass Pieces, 30 Polyethylene Pieces, 30 Rubber Stoppers, 30 Balloons, 15 Push Pins, 15 Plastic Cups, 20 Cardboard Pieces, 2 Pk. of Toothpicks, 150 Plastic Stirrers, 2 Pk. of Sticky Connecting Putty, 150 Small Wooden Skewers, 100 Rubber Bands, 100 Straws, 100 Wooden Craft Sticks, 100 Index Cards, 2 Pk. of String, Teacher Manual, and Student Guide copy masters
Material Cardboard Box
Quantity 1 Kit
Weight (English) ∼7 lb.
Weight (Metric) ∼3.18 kg
Content And Storage Ambient
Product Line Innovating Science
Dimensions (L x W x H) 28.75 x 16 x 5 in.
Class Size 15 Lab Groups
Product Type Engineer and Explore Materials for Prosthetics Kit
Show More Show Less

WARNING: This product can expose you to chemicals including Phthalates, which is known to the State of California to cause reproductive harm. For more information go to www.P65Warnings.ca.gov.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.