Learn More
ENH/PDLIM5 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$167.95
Specifications
| Antigen | ENH/PDLIM5 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Liquid |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-1028
|
Bethyl Laboratories, Inc
A301704AT |
20 μL |
Each of 1 for $167.95
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat
May play an important role in the heart development by scaffolding PKC to the Z-disk region. May play a role in the regulation of cardiomyocyte expansion. Overexpression promotes the development of heart hypertrophy. Contributes to the regulation of dendritic spine morphogenesis in neurons. May restrain postsynaptic growth of excitatory synapses.Specifications
| ENH/PDLIM5 | |
| Polyclonal | |
| Liquid | |
| RUO | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| ENH, ENH1, LIM, PDZ and LIM domain protein 5, enigma homolog, enigma-like LIM domain protein, enigma-like PDZ and LIM domains protein | |
| Between 546 and 596 | |
| Primary | |
| 4° C | |
| PDLIM5 |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| Q8CI51, Q96HC4 | |
| 10611, 56376 | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.