Learn More
ENH/PDLIM5 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301703AT
Description
The recommended shelf life for this product is 1 year from date of receipt.
May play an important role in the heart development by scaffolding PKC to the Z-disk region. May play a role in the regulation of cardiomyocyte expansion. Overexpression promotes the development of heart hypertrophy. Contributes to the regulation of dendritic spine morphogenesis in neurons. May restrain postsynaptic growth of excitatory synapses.Specifications
| ENH/PDLIM5 | |
| Polyclonal | |
| Unconjugated | |
| PDLIM5 | |
| ENH, ENH1, LIM, PDZ and LIM domain protein 5, enigma homolog, enigma-like LIM domain protein, enigma-like PDZ and LIM domains protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10611 | |
| 4° C | |
| Liquid |
| Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q96HC4 | |
| PDLIM5 | |
| Between 375 and 425 | |
| 10 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.