Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Minigrip™ Enhanced SPECI-GARD™ Biohazard Bags
Click to view available options
Color:
Clear
White
Description
- Separate document pocket with flap
- Press and Close™ secure sample closure provides infection prevention and user safety
- Utilizes “no glove to adhesive” contact capability when sealing bags
- Easy-to-open tear line
- Simple instructions for use printed on bags
Order Info
All bags measure 6W x 10 in. H.
Specifications
Specifications
Color | Clear |
For Use With (Application) | Easy-to-open tear line |
Includes | Adhesive closure, 2 pockets, biohazard label |
Material | Polyethylene |
Width (English) | 6 in. |
Width (Metric) | 15.24 cm |
Height (English) | 10 in. |
Height (Metric) | 25.4 cm |
Product Type | Biohazard Bag |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction