Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
enkurin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | enkurin |
---|---|
Dilution | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
enkurin Polyclonal specifically detects enkurin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
enkurin | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
C10orf63DKFZp781F21103, chromosome 10 open reading frame 63, enkurin, enkurin, TRPC channel interacting protein, MGC26778 | |
ENKUR | |
IgG | |
Affinity Purified |
Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Rabbit | |
Human | |
Q8TC29 | |
219670 | |
This antibody was developed against a recombinant protein corresponding to amino acids: SSECIYNLIPSDLKEPPQPPRYISIFKATVKDDMQKAKTAMKTMGPAKVEVPSPKDFLKKHSKEKTLPPKKNFDRNVP | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title