Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
enkurin Antibody (OTI6C10), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272461UV
Description
enkurin Monoclonal specifically detects enkurin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
enkurin | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ENKUR | |
Full length recombinant protein of human enkurin (NP_659447) produced in E.coli. | |
0.1 mL | |
219670.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI6C10 | |
Western Blot, Immunohistochemistry | |
C10orf63DKFZp781F21103, chromosome 10 open reading frame 63, enkurin, enkurin, TRPC channel interacting protein, MGC26778 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction