Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
enkurin Antibody (SR1480), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | enkurin |
---|---|
Clone | SR1480 |
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
enkurin Monoclonal antibody specifically detects enkurin in Human, Mouse samples. It is validated for Western BlotSpecifications
enkurin | |
Western Blot 1:500-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
219670 | |
IgG | |
Affinity purified |
SR1480 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
C10orf63DKFZp781F21103, chromosome 10 open reading frame 63, enkurin, enkurin, TRPC channel interacting protein, MGC26778 | |
A synthesized peptide derived from human enkurin (Uniprot #: Q8TC29) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title