Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Enkurin Monoclonal Antibody (OTI6C10)
GREENER_CHOICE

Catalog No. PIMA526653
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526653 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526653 Supplier Invitrogen™ Supplier No. MA526653
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Enkurin Monoclonal Antibody for Western Blot, IHC (P)

Enkurin interacts with transient receptor potential canonical.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Enkurin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI6C10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ENKUR
Gene Accession No. Q8TC29
Gene Alias 4933434I06Rik; AU016229; C10orf63; CFAP106; Enkur; Enkurin; enkurin, TRPC channel interacting protein; RGD1309839
Gene Symbols ENKUR
Host Species Mouse
Immunogen Full length human recombinant protein of ENKUR produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219670
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.