Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Enkurin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563969
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563969 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563969 Supplier Invitrogen™ Supplier No. PA563969
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SSECIYNLIP SDLKEPPQPP RYISIFKATV KDDMQKAKTA MKTMGPAKVE VPSPKDFLKK HSKEKTLPPK KNFDRNVP Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 77%.

Enkurin interacts with transient receptor potential canonical.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Enkurin
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ENKUR
Gene Accession No. Q8TC29
Gene Alias 4933434I06Rik; AU016229; C10orf63; CFAP106; Enkur; Enkurin; enkurin, TRPC channel interacting protein; RGD1309839
Gene Symbols ENKUR
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Enkurin. Recombinant protein control fragment (Product #RP-103408).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219670
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.