Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Enkurin Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | Enkurin |
---|---|
Concentration | 0.28 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-5339
![]() ![]() |
Proteintech Group Inc
264401AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Enkurin interacts with transient receptor potential canonical.Specifications
Enkurin | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q6SP97, Q8TC29 | |
219670, 71233 | |
Enkurin Fusion Protein Ag23762 | |
Primary | |
-20°C | |
ENKUR |
0.28 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
C10orf63, ENKUR, Enkurin | |
Enkur | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title