Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENO1 Monoclonal Antibody (4D11F5), Invitrogen™

Catalog No. FEMA550266
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550266 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550266 Supplier Thermo Scientific Supplier No. MA550266

Mouse Monoclonal Antibody

Enolase (2-phosphogly-cerate hydrolyase or phosphopyruvate hydrates) is a glycolytic enzyme that catalyzes the dehydration and conversion of 2-phosphoglycerate to phosphoenolpyruvate. It comprises three distinct subunits, alpha, beta and gamma. Non Neuronal Enolase(ENO1) is an isoform of mammalian enolase and is composed of 2 alpha subunits. The gene for ENO1 also encodes a shorter monomeric structural lens protein, tau-crystallin. Multifunctional enzyme that, as well as its role in glycolysis, plays a part in various processes such as growth control, hypoxia tolerance and allergic responses. May also function in the intravascular and pericellular fibrinolytic system due to its ability to serve as a receptor and activator of plasminogen on the cell surface of several cell-types such as leukocytes and neurons. Stimulates immunoglobulin production.Used as a diagnostic marker for many tumors and, in the heterodimeric form, alpha/gamma, as a marker for hypoxic brain injury after cardiac arrest. Also marker for endometriosis. Antibodies against alpha-enolase are present in sera from patients with cancer-associated retinopathy syndrome (CAR), a progressive blinding disease which occurs in the presence of systemic tumor growth, primarily small-cell carcinoma of the lung and other malignancies. Is identified as an autoantigen in Hashimoto encephalopathy (HE) a rare autoimmune disease associated with Hashimoto thyroiditis (HT). HT is a disorder in which destructive processes overcome the potential capacity of thyroid replacement leading to hypothyroidism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ENO1
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 4D11F5
Concentration 2.765 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300, pH 7.4
Gene ENO1
Gene Accession No. P04764, P06733, P17182
Gene Alias 0610008I15, 2-phospho-D-glycerate hydrolase, 2-phospho-D-glycerate hydro-lyase, AL022784, alpha enolase, alpha enolase like 1, alpha-enolase, alpha-enolase, c-myc promoter-binding protein-1, c-Myc promoter binding protein, C-myc promoter-binding protein, Eno1, Eno-1, ENO1L1, enoa, Enolase 1, enolase 1, (alpha), enolase 1, alpha non-neuron, enolase-alpha, epididymis secretory protein Li 17, HEL-S-17, MBP-1, MBPB1, MPB1, MPB-1, MYC promoter-binding protein 1, NNE, Non-neural enolase, phosphopyruvate hydratase, Plasminogen-binding protein, PPH, tau-crystallin
Gene Symbols ENO1
Host Species Mouse
Immunogen Recombinant Human Alpha-enolase protein (2-434AA)
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100352426, 13806, 2023, 24333
Target Species Human, Mouse, Rabbit, Rat
Content And Storage -20°C or -80°C if preferred
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.