Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ENO1 Polyclonal Antibody
ENO1 Polyclonal Antibody
Supplier: Thermo Scientific PA533143
Description
ENO1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (F), IHC (P), ELISA

Specifications
ENO1 | |
Polyclonal | |
Unconjugated | |
ENO1 | |
0610008I15; 2-phospho-D-glycerate hydrolase; 2-phospho-D-glycerate hydro-lyase; AL022784; alpha enolase; alpha enolase like 1; alpha-enolase; alpha-enolase; c-myc promoter-binding protein-1; c-Myc promoter binding protein; C-myc promoter-binding protein; Eno1; Eno-1; ENO1L1; enoa; Enolase 1; enolase 1, (alpha); enolase 1, alpha non-neuron; enolase-alpha; epididymis secretory protein Li 17; HEL-S-17; MBP-1; MBPB1; MPB1; MPB-1; MYC promoter-binding protein 1; NNE; Non-neural enolase; phosphopyruvate hydratase; Plasminogen-binding protein; PPH; tau-crystallin | |
Rabbit | |
Protein G | |
RUO | |
2023 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
5 mg/mL | |
PBS with 0.09% sodium azide | |
P06733 | |
ENO1 | |
Native human NNE. | |
1 mL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction